Al-23 Fish Spine Beads;OD (mm), 3.5;Bore (mm), 1.0;Length (mm), 3.0
Thermo Scientific Chemicals
Al-23 Fish Spine Beads;OD (mm), 3.5;Bore (mm), 1.0;Length (mm), 3.0
Have Questions?
Catalog NumberQuantity
032905.KX100 Pk.
Catalog number 032905.KX
Price (USD)
150.65
Special offer
Online exclusive
Ends: 30-Jun-2026
177.00
Save 26.35 (15%)
Each
Quantity:
100 Pk.
Request bulk or custom format
Price (USD)
150.65
Special offer
Online exclusive
Ends: 30-Jun-2026
177.00
Save 26.35 (15%)
Each
Specifications
Chemical Name or MaterialAl-23 Fish Spine Beads
Recommended StorageAmbient temperatures
TSCAU
FormFish Spine Beads
Quantity100 Pk.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Frequently asked questions (FAQs)
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.