Al-23 Fish Spine Beads;OD (mm), 4.0;Bore (mm), 1.5;Length (mm), 4.7 - FAQs
View additional product information for Al-23 Fish Spine Beads;OD (mm), 4.0;Bore (mm), 1.5;Length (mm), 4.7 - FAQs (032906.KX)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.