Search
View additional product information for Al-23 Fish Spine Beads;OD (mm), 7.0;Bore (mm), 3.8;Length (mm), 7.0 - FAQs (032908.KX)
1 product FAQs found
The maximum working temperature for the Al-23 Tube is 1950 degrees C.