Cadmium bromide tetrahydrate, Reagent Grade
Cadmium bromide tetrahydrate, Reagent Grade
Cadmium bromide tetrahydrate, Reagent Grade
Thermo Scientific Chemicals

Cadmium bromide tetrahydrate, Reagent Grade

CAS: 13464-92-1 | Br2CdH8O4 | 344.282 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 032958.14
also known as 032958-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS13464-92-1
IUPAC Namecadmium(2+) tetrahydrate dibromide
Molecular FormulaBr2CdH8O4
InChI KeyPANYFCXNZSEXSU-UHFFFAOYSA-L
SMILESO.O.O.O.[Br-].[Br-].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content1% max
It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only