Al-23 Crucible, Conical;Ht (mm), 30;Top OD (mm), 25;Top ID (mm), 21;Base OD (mm), 13;Volume (ml), 9 - FAQs
View additional product information for Al-23 Crucible, Conical;Ht (mm), 45;Top OD (mm), 35;Top ID (mm), 31;Base OD (mm), 18;Volume (ml), 23 - FAQs (033010.KF)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.