4-Methyl-1-pentene, 98+%
4-Methyl-1-pentene, 98+%
4-Methyl-1-pentene, 98+%
4-Methyl-1-pentene, 98+%
Thermo Scientific Chemicals

4-Methyl-1-pentene, 98+%

CAS: 691-37-2 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 033029.30
also known as 033029-30
Price (USD)
714.65
Online Exclusive
794.00
Save 79.35 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
714.65
Online Exclusive
794.00
Save 79.35 (10%)
Each
Add to cart
Specifications
Beilstein1731096
CAS691-37-2
Chemical Name or Material4-Methyl-1-pentene
ColorColorless
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
View more
4-Methyl-1-pentene is used as a monomer for olefin polymerisation. The resulting polymer is poly(4-methyl-1-pentene). It can also be used to produce 1,2-diiodo-4-methyl-pentane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only