Al-23 Round Disc Without Hole;OD (mm), 10;Thick (mm), 3 - FAQs
View additional product information for Al-23 Round Disc Without Hole;OD (mm), 10;Thick (mm), 3 - FAQs (033107.KF)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.