What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.
Search
View additional product information for Al-23 Boat;L1 (mm), 48;L2 (mm), 46;B1 (mm), 9;B2 (mm), 8;H (mm), 6;W (mm), 2;Vol, 0.6 - FAQs (033171.KF)
1 product FAQs found