Al-23 Boat;L1 (mm), 87;L2 (mm), 72;B1 (mm), 15;B2 (mm), 13;H (mm), 8;W (mm), 2.5;Vol, 3.4 - FAQs
Al-23 Boat;L1 (mm), 87;L2 (mm), 72;B1 (mm), 15;B2 (mm), 13;H (mm), 8;W (mm), 2.5;Vol, 3.4 - FAQs
View additional product information for Al-23 Boat;L1 (mm), 87;L2 (mm), 72;B1 (mm), 15;B2 (mm), 13;H (mm), 8;W (mm), 2.5;Vol, 3.4 - FAQs (033175.KF)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.