Al-23 Boat;L1 (mm), 130;L2 (mm), 116;B1 (mm), 31;B2 (mm), 27;H (mm), 17;W (mm), 3;Vol, 36.0 - FAQs
View additional product information for Al-23 Boat;L1 (mm), 130;L2 (mm), 116;B1 (mm), 31;B2 (mm), 27;H (mm), 17;W (mm), 3;Vol, 36.0 - FAQs (033182.KF)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.