Al-23 Combustion Tray;L1 (mm), 100;L2 (mm), 98;B1 (mm), 50;B2 (mm), 48;H (mm), 30;W (mm), 4;Vol, 100.0
Thermo Scientific Chemicals
Al-23 Combustion Tray;L1 (mm), 100;L2 (mm), 98;B1 (mm), 50;B2 (mm), 48;H (mm), 30;W (mm), 4;Vol, 100.0
Have Questions?
Catalog NumberQuantity
033186.KF1 Ea.
Catalog number 033186.KF
Price (USD)
252.65
Online Exclusive
280.00
Save 27.35 (10%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
252.65
Online Exclusive
280.00
Save 27.35 (10%)
Each
Specifications
Chemical Name or MaterialAl-23 Combustion Tray
Recommended StorageAmbient temperatures
TSCAU
Quantity1 Ea.
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Frequently asked questions (FAQs)
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.