Al-24 Combustion Tray;L (mm), 145;L2 (mm), 138;B (mm), 85;B2 (mm), 80;H (mm), 30;W (mm), 5;Vol, 240 - FAQs
View additional product information for Al-24 Combustion Tray;L (mm), 145;L2 (mm), 138;B (mm), 85;B2 (mm), 80;H (mm), 30;W (mm), 5;Vol, 240 - FAQs (033187.KF)
1 product FAQs found
What is the maximum working temperature of Al-24 Tubes?
The maximum working temperature for the Al-24 Tube is 1950 degrees C.