Aluminum oxide, fused, insulating powder, 99.7+%
Aluminum oxide, fused, insulating powder, 99.7+%
Aluminum oxide, fused, insulating powder, 99.7+%
Thermo Scientific Chemicals

Aluminum oxide, fused, insulating powder, 99.7+%

Used in Dry Filling For thermal Insulation of Electrical Furnaces | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 033199.A1
also known as 033199-A1
Price (USD)
87.65
Online exclusive
97.30
Save 9.65
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
87.65
Online exclusive
97.30
Save 9.65
Each
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.70% Al₂O₃
Used in dry filling for thermal insulation of electrical furnaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Form: approx. 180 micron
RUO – Research Use Only