Aluminum oxide, cement, Al2O3 95% (SiO2 ∽5%), Thermo Scientific Chemicals
Aluminum oxide, cement, Al2O3 95% (SiO2 ∽5%), Thermo Scientific Chemicals
Aluminum oxide, cement, Al2O3 95% (SiO2 ∽5%), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum oxide, cement, Al2O3 95% (SiO2 ∽5%), Thermo Scientific Chemicals

Form: approx. 60 micron
Have Questions?
Catalog NumberQuantity
033201.A1
also known as 033201-A1
1 kg
Catalog number 033201.A1
also known as 033201-A1
Price (USD)
82.65
Online Exclusive
91.40
Save 8.75 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
82.65
Online Exclusive
91.40
Save 8.75 (10%)
Each
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofA≥90% alumina, Al2O3
CommentSuitable for use as cement
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What are the recommended guidelines for use of the Aluminum oxide, cement, Al2O3 95% (SiO2 ∽5%) (Cat. No. 033201.A1)?

We recommend the following guidelines for use of Aluminum oxide, cement, Al2O3 95% (SiO2 ~5%) (Cat. No. 033201.A1):
Mixing: Add a minimum amount of water until the cement has reach your desired consistency.
Hardening: This product it is not cold-hardening. It contains an organic binder to increase dry strength. The organic binder is completely burned off during the heating process. Above 1200 degrees C, progressive hardening of the cement will occur as a result of sintering.
Curing: The curing time can vary depending on several factors, including: the amount of water used to form the slurry, the temperature used during hardening, and the degree of sintering desired (if heating to temperatures over 1200 degrees C).