Phenol, ACS, 99+%, stab.
Phenol, ACS, 99+%, stab.
Phenol, ACS, 99+%, stab.
Thermo Scientific™

Phenol, ACS, 99+%, stab.

CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 033213.22
also known as 033213-22
Price (USD)
52.40
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
52.40
Each
Chemical Identifiers
CAS108-95-2
IUPAC Namephenol
Molecular FormulaC6H6O
InChI KeyISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Iodometric Titration)≥99.0%
Appearance (Color)Colorless
FormCryslals or crystalline solid
CommentMay become pink on exposure to air
Freezing PointNot below 40.5°C (dry basis)
View more
Phenol is used as a precursor for cyclohexanone, plastics, nonionic detergents and pharmaceutical drugs like aspirin. It acts as an anesthetic in chloraseptic. It reacts with acetone to get bisphenol-A which is used in the synthesis of poly carbonates and epoxide resins. It is also used in the manufacture of synthetic resins, dyes, pharmaceuticals, synthetic tanning agents, perfumes, lubricating oils and solvents. In molecular biology, it is used in the extraction of nucleic acid from tissues by using the liquid/liquid phenol-chloroform extraction technique. It is an active component of paint strippers which are used for the removal of epoxy and polyurethane. It is also used in the preparation of cosmetics, hair colors and skin lightening preparations. In the field of medicine, its spray is useful in helping sore throat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is used for a stabilizer in Phenol, ACS, 99+%, stab.?

The stabilizer in Phenol, ACS, 99+%, stab. is hypophosphorous acid. It is included at a maximum concentration of 0.15%.