Bismuth Lead Tin Cadmium ingot (Wood's metal)
Bismuth Lead Tin Cadmium ingot (Wood's metal)
Bismuth Lead Tin Cadmium ingot (Wood's metal)
Thermo Scientific Chemicals

Bismuth Lead Tin Cadmium ingot (Wood's metal)

CAS: 8049-22-7 | Cd12Sn2 | 1586.388 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 033218.36
also known as 033218-36
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS8049-22-7
IUPAC Namedodecacadmium ditin
Molecular FormulaCd12Sn2
InChI KeyOIBLPQFTAJDRMC-UHFFFAOYSA-N
SMILES[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Sn].[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
FormIngot
CompositionBismuth (Bi): 49.0-51.0%
CompositionCadmium (Cd): 12.0-13.0%
CompositionLead (Pb): 24.0-26.0%
CompositionTin (Sn): 12.0-13.0%
Wood's metal is useful as a low-melting solder, low-temperature casting metal, high temperature coupling fluid in heat baths, and as a fire-melted valve element in fire sprinkler systems in buildings. It is also used as a filler when bending thin-walled metal tube.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only