Thermo Scientific Chemicals

Sodium bismuth oxide, ACS, 80% min, Thermo Scientific Chemicals

Catalog number: 033219.22
100 g, Each
Thermo Scientific Chemicals

Sodium bismuth oxide, ACS, 80% min, Thermo Scientific Chemicals

Catalog number: 033219.22
100 g, Each
Quantity
Catalog number: 033219.22
also known as 033219-22
Price (USD)
Price: 235.00
Online price: 199.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12232-99-4
IUPAC Name
sodium bismuthanoylolate
Molecular Formula
BiNaO3
InChI Key
PNYYBUOBTVHFDN-UHFFFAOYSA-N
SMILES
[Na+].[O-][Bi](=O)=O
Impurity content
Oxidizing efficiency 99.6% min
Impurity content
Cl 0.002%
Impurity content
Mn 5ppm

Description

Sodium bismuth oxide is used for the determination of manganese in iron, steel and ores. It is also used in the separation of plutonium and acts as a strong oxidizer. It is used in the conversion of manganese compound to permanganate. Further, it is used in the preparation of hexavalent americium (Am) during its separation from the lanthanides and in the hydrothermal synthesis of bismuth oxychloride micrometer-sized single crystals. In addition to this, it is employed as a precursor in the preparation of bismuth nanoforms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text