Potassium sodium L-tartrate tetrahydrate, ACS, 99.0-102.0%
Potassium sodium L-tartrate tetrahydrate, ACS, 99.0-102.0%
Potassium sodium L-tartrate tetrahydrate, ACS, 99.0-102.0%
Thermo Scientific Chemicals

Potassium sodium L-tartrate tetrahydrate, ACS, 99.0-102.0%

CAS: 6381-59-5 | C4H12KNaO10 | 282.218 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 033241.A4
also known as 033241-A4
Price (USD)
401.65
Online Exclusive
446.00
Save 44.35 (10%)
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
401.65
Online Exclusive
446.00
Save 44.35 (10%)
Each
Chemical Identifiers
CAS6381-59-5
IUPAC Namepotassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
Molecular FormulaC4H12KNaO10
InChI KeyVZOPRCCTKLAGPN-ZFJVMAEJSA-L
SMILESO.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentChloride (as Cl) 0.0015
Impurity contentFe 0.001%
Impurity contentPO₄ 0.002%
Impurity contentSO₄ 0.005%
Impurity contentCa 0.005%
View more
Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is an active ingredient in Fehiling’s solution and Nylander’s test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as a laxative and is also used in food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only