Oxalic acid dihydrate, ACS, 99.5-102.5%
Oxalic acid dihydrate, ACS, 99.5-102.5%
Oxalic acid dihydrate, ACS, 99.5-102.5%
Oxalic acid dihydrate, ACS, 99.5-102.5%
Thermo Scientific Chemicals

Oxalic acid dihydrate, ACS, 99.5-102.5%

CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
5 kg
Catalog number 033262.18
also known as 033262-18
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
40.60
Save 5.95 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
40.60
Save 5.95 (15%)
Each
Chemical Identifiers
CAS6153-56-6
IUPAC Nameoxalic acid dihydrate
Molecular FormulaC2H6O6
InChI KeyGEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILESO.O.OC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or colorless
FormCrystalline powder or solid
Assay (Redox Titration)99.5-102.5%
Calcium content≤0.001%
Chloride content≤0.002%
View more
As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing and as a reagent for dehydration and condensation Oxalic acid dihydrate acts as an analytical reagent, as a reducing agent for photographic films and as mordant in the dyeing process. It is used as a plating agent, a surface treating agent, a grinding agent, a precipitating agent in rare-earth mineral processing, a bleaching agent in textile and a purifying agent in the pharmaceutical industry. It plays an important role in antibiotic medication like oxytetracycline and chloramphenicol. It finds application in water treatment plants to remove calcium waste water. It is also used as a buffer in chromatographic separation as well as bleach for wood, removing black stains caused by water penetration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing, and as a reagent for dehydration and condensation
RUO – Research Use Only