Cyclohexanone, ACS, 99+%
Cyclohexanone, ACS, 99+%
Cyclohexanone, ACS, 99+%
Thermo Scientific Chemicals

Cyclohexanone, ACS, 99+%

CAS: 108-94-1 | C6H10O | 98.145 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
4 x 1 L
Catalog number 033309.K7
also known as 033309-K7
Price (USD)
154.65
Special offer
Online exclusive
Ends: 30-Jun-2026
182.00
Save 27.35 (15%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
154.65
Special offer
Online exclusive
Ends: 30-Jun-2026
182.00
Save 27.35 (15%)
Each
Chemical Identifiers
CAS108-94-1
IUPAC Namecyclohexanone
Molecular FormulaC6H10O
InChI KeyJHIVVAPYMSGYDF-UHFFFAOYSA-N
SMILESO=C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤10
Assay (GC)≥99.0%
Residual water≤0.05%
Residue on Evaporation≤0.05%
Cyclohexanone is used as an industrial solvent and paint remover. It acts as a precursor to nylon 6,6 and nylon 6 and cyclohexanone oxime, which gives caprolactam on rearrangement. Further, it is used as a chemical reaction medium, adhesives, sealants and agricultural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only