Iron(III) sulfate hydrate, Reagent Grade
Iron(III) sulfate hydrate, Reagent Grade
Iron(III) sulfate hydrate, Reagent Grade
Thermo Scientific Chemicals

Iron(III) sulfate hydrate, Reagent Grade

CAS: 15244-10-7 | Fe2H2O13S3 | 417.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 033316.30
also known as 033316-30
Price (USD)
59.00
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
59.00
Each
Chemical Identifiers
CAS15244-10-7
IUPAC Namediiron(3+) hydrate trisulfate
Molecular FormulaFe2H2O13S3
InChI KeyVXWSFRMTBJZULV-UHFFFAOYSA-H
SMILESO.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey or pale yellow to yellow
FormPowder or crystalline powder
Assay (unspecified)73% Fe₂(SO₄)₃ min
Iron(III) sulfate is also used in pigments, pickling baths for aluminum and steel. It acts as a mordant in dyeing, as a coagulant for industrial wastes. In the medical field, it is used as an astringent and styptic. It is involved in the preparation of aromatic esters from the corresponding acids and alcohols. Further, it is employed in the oxidation of pyrite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only