Hexanes, mixed isomers, ACS, 98.5+%
Hexanes, mixed isomers, ACS, 98.5+%
Hexanes, mixed isomers, ACS, 98.5+%
Thermo Scientific Chemicals

Hexanes, mixed isomers, ACS, 98.5+%

CAS: 92112-69-1 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
4 L
4 x 1 L
Catalog number 033321.K2
also known as 033321-K2
Price (USD)
63.80
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
63.80
Each
Chemical Identifiers
CAS92112-69-1
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)(Water soluble) ≤0.0003 meq/g
FormLiquid
Color (APHA)≤10
Assay (GC)≥98.5% hexanes ( sum of 5 isomers, total hexanes, plus methylcyclopentane
Impurity contentSulfur compounds (as S) ≤0.005%
View more
Hexane mixed isomers is used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds namely butyl lithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only