Manganese(II) sulfate monohydrate, ACS, 98.0-101.0%
Manganese(II) sulfate monohydrate, ACS, 98.0-101.0%
Manganese(II) sulfate monohydrate, ACS, 98.0-101.0%
Manganese(II) sulfate monohydrate, ACS, 98.0-101.0%
Thermo Scientific Chemicals

Manganese(II) sulfate monohydrate, ACS, 98.0-101.0%

CAS: 10034-96-5 | H2MnO5S | 169.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 033341.36
also known as 033341-36
Price (USD)
67.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
67.00
Each
Add to cart
Chemical Identifiers
CAS10034-96-5
IUPAC Namemanganese(2+) hydrate sulfate
Molecular FormulaH2MnO5S
InChI KeyISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILESO.[Mn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0-101.0% MnSO₄?H₂O
Impurity contentCa 0.005%
Impurity contentK 0.01%
Impurity contentSubstances reducing permanganate P.T.
Insoluble material0.01%
View more
Manganese(II) sulfate monohydrate is used as a colorant in dyes, fertilizers, animal feeds and red glazes on porcelain. Further, it is used in paints, ceramics, nutrient and dietary supplement. It is involved in the preparation of manganese dioxide. In addition, it serves as a precursor to manganese metal and other manganese compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only