Ethanol, Alcohol Reagent, anhydrous, denatured, ACS, 94-96%
Ethanol, Alcohol Reagent, anhydrous, denatured, ACS, 94-96%
Ethanol, Alcohol Reagent, anhydrous, denatured, ACS, 94-96%
Thermo Scientific Chemicals

Ethanol, Alcohol Reagent, anhydrous, denatured, ACS, 94-96%

CAS: 64-17-5 | C2H6O | 46.069 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
4 L
4 x 1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 033361.M1
also known as 033361-M1
Price (USD)
-
Quantity:
4 x 1 L
Request bulk or custom format
Chemical Identifiers
CAS64-17-5
IUPAC Nameethanol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)Within 94.0-96.0% (V/V) methanol and ethyl alcohol
Assay (GC)Isopropyl alcohol (2-propanol)(v/v) 4.0-6.0%
Water Content (Karl Fischer Titration)≤0.5%
Residue on Evaporation≤0.001%
View more
Ethanol is used in antibacterial hand sanitizer gels, as an antidote to methanol poisoning and an antitussive agent. It is acting as a solvent to remove the paint and ink stain on curtains, dress and upholstery. It is utilized as a fuel in camping and primus stoves. It is likewise employed to clean the windows, manufacturing nail-polish and to test pectin levels in jam and jelly making. It finds application in personal care products such as mouthwashes, perfumes and deodorants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use