Sodium perborate tetrahydrate, Reagent Grade
Sodium perborate tetrahydrate, Reagent Grade
Sodium perborate tetrahydrate, Reagent Grade
Sodium perborate tetrahydrate, Reagent Grade
Thermo Scientific Chemicals

Sodium perborate tetrahydrate, Reagent Grade

CAS: 10486-00-7 | NaBO3·4H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 033383.22
also known as 033383-22
Price (USD)
46.10
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
46.10
Each
Chemical Identifiers
CAS10486-00-7
IUPAC Namedisodium tetrahydroxy-1,2,4,5,3,6-tetraoxadiborinane-3,6-diuide hexahydrate
Molecular FormulaB2H16Na2O14
InChI KeyMCYPYXIRGFEFGL-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Na+].[Na+].O[B-]1(O)OO[B-](O)(O)OO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)NaBO₃?4H₂O 96% min
Sodium perborate tetrahydrate is mainly used in textile mills as a bleaching agent for a variety of fabrics, namely, wool, cotton, linen, rayon, etc, and as oxidizing and fixing agent for dyes. It is also used in dentifrices, denture cleaners and pharmaceuticals due to its cleansing, deodorizing and antiseptic properties and is incorporated in soaps and detergent powders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Mortar, plasters, cements, gas manufacturing to remove ammonia, in lubricants, water softening, petrochemicals, soil conditioner, paving materials
RUO – Research Use Only