Sodium peroxide, ACS, 93% min
Sodium peroxide, ACS, 93% min
Sodium peroxide, ACS, 93% min
Thermo Scientific Chemicals

Sodium peroxide, ACS, 93% min

For Use As An Oxidizing Agent, a Bleaching Agent, Deodorant | CAS: 1313-60-6 | Na2O2 | 77.978 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Catalog number 033384.09
also known as 033384-09
Price (USD)
26.50
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
26.50
Each
Chemical Identifiers
CAS1313-60-6
IUPAC Namedisodium dioxidanediide
Molecular FormulaNa2O2
InChI KeyPFUVRDFDKPNGAV-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy Metals (as Pb)≤20ppm
Impurity contentIron, Fe: ≤50ppm
Impurity contentPhosphate PO₄: ≤5ppm
Impurity contentSulfate SO₄: ≤10ppm
FormPowder and granules
View more
As an oxidizing agent, a bleaching agent, deodorant, pharmaceuticals and dyes, and as an analytical reagent.Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only