Strontium chloride hexahydrate, ACS, 99.0-103.0%
Strontium chloride hexahydrate, ACS, 99.0-103.0%
Strontium chloride hexahydrate, ACS, 99.0-103.0%
Thermo Scientific Chemicals

Strontium chloride hexahydrate, ACS, 99.0-103.0%

CAS: 10025-70-4 | Cl2H12O6Sr | 266.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 033393.36
also known as 033393-36
Price (USD)
134.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
134.00
Each
Chemical Identifiers
CAS10025-70-4
IUPAC Namestrontium(2+) hexahydrate dichloride
Molecular FormulaCl2H12O6Sr
InChI KeyAMGRXJSJSONEEG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentBa 0.05%
Impurity contentMg 2ppm
Assay (unspecified)99.0-103.0% SrCl₂.6H₂O
Heavy Metals(ICP-OES) 5ppm
Impurity contentCa 0.05%
View more
Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the medical field, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only