Sulfur powder, sublimed, -100 mesh, 99.5%
Sulfur powder, sublimed, -100 mesh, 99.5%
Sulfur powder, sublimed, -100 mesh, 99.5%
Sulfur powder, sublimed, -100 mesh, 99.5%
Thermo Scientific Chemicals

Sulfur powder, sublimed, -100 mesh, 99.5%

CAS: 7704-34-9 | S | 32.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 033394.A3
also known as 033394-A3
Price (USD)
148.65
Online Exclusive
165.00
Save 16.35 (10%)
Each
Quantity:
2 kg
Request bulk or custom format
Price (USD)
148.65
Online Exclusive
165.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS7704-34-9
IUPAC Namesulfur
Molecular FormulaS
InChI KeyNINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES[S]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.5% minimum
Particle Size-100 mesh (less than 150 micron)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only