Zinc sulfate heptahydrate, ACS, 99.0-103.0%
Zinc sulfate heptahydrate, ACS, 99.0-103.0%
Zinc sulfate heptahydrate, ACS, 99.0-103.0%
Zinc sulfate heptahydrate, ACS, 99.0-103.0%
Thermo Scientific Chemicals

Zinc sulfate heptahydrate, ACS, 99.0-103.0%

CAS: 7446-20-0 | H14O11SZn | 287.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 033399.36
Price (USD)
82.70
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
82.70
Each
Chemical Identifiers
CAS7446-20-0
IUPAC Namezinc(2+) heptahydrate sulfate
Molecular FormulaH14O11SZn
InChI KeyRZLVQBNCHSJZPX-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0-103.0% O₄SZn.7H₂O
Impurity contentFe 0.001%
Impurity contentMg 0.005%
Impurity contentPb 0.003%
Impurity contentCa 0.005%
View more

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only