Sodium hydroxide, 25% in aqueous solution, Biotech Reagent, GMP, J.T.Baker™
Avantor™

Sodium hydroxide, 25% in aqueous solution, Biotech Reagent, GMP, J.T.Baker™

Made from USP purified water and multicompendial sodium hydroxide pellets. Filtered through a 0.2 μm element. GMP manufactured product.
Have Questions?
Change viewbuttonViewtableView
Quantity:
19 L
1135 L
Catalog number 0338-07
Price (USD)
-
Quantity:
19 L
Made from USP purified water and multicompendial sodium hydroxide pellets. Filtered through a 0.2 μm element. GMP manufactured product.
Made from USP purified water and multicompendial sodium hydroxide pellets. Filtered through a 0.2 μm element. GMP manufactured product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point119°C to 135°C (1013 hPa)
CAS1310-73-2
Chemical Name or MaterialSodium hydroxide
ColorColorless
Melting Point9°C to 10°C
Molecular FormulaNaOH
PackagingPoly bottle, corrugated shipper
SynonymSodium hydrate, Soda lye, Caustic soda, Lye
GradeBiotech
Molecular Weight (g/mol)40
Quantity19 L
Unit SizeEach