Diethylgermanium dichloride
Diethylgermanium dichloride
Diethylgermanium dichloride
Thermo Scientific Chemicals

Diethylgermanium dichloride

As an intermediate | CAS: 13314-52-8 | C4H14Cl2Ge | 205.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 034108.14
also known as 034108-14
Price (USD)
1,331.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,567.00
Save 235.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
1,331.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,567.00
Save 235.35 (15%)
Each
Specifications
CAS13314-52-8
Chemical Name or MaterialDiethylgermanium dichloride
ColorColorless
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Density1.372 g/mL
View more
As an intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only