Tetra-n-butylgermanium, 99%
Tetra-n-butylgermanium, 99%
Tetra-n-butylgermanium, 99%
Thermo Scientific Chemicals

Tetra-n-butylgermanium, 99%

CAS: 1067-42-1 | C16H36Ge | 301.094 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 034119.06
also known as 034119-06
Price (USD)
145.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
145.00
Each
Specifications
CAS1067-42-1
Chemical Name or MaterialTetra-n-butylgermanium
Density0.934 g/mL at 20°C
EINECS Number213-929-3
Flash Point>110°C (230°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only