What concentrations of Pb (lead) can the Lead, Quant Test Strips test for?
The Lead, Quant Test Strips can measure concentrations of 20-500 ppm Pb+2 in aqueous solutions.
Search
View additional product information for Lead, Quant™ Test Strips - FAQs (034933.QA)
2 product FAQs found