Niobium slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, Puratronic™, 99.99% (metals basis excluding Ta), Ta 300ppm
Niobium slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, Puratronic™, 99.99% (metals basis excluding Ta), Ta 300ppm
Niobium slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, Puratronic™, 99.99% (metals basis excluding Ta), Ta 300ppm
Thermo Scientific Chemicals

Niobium slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, Puratronic™, 99.99% (metals basis excluding Ta), Ta 300ppm

CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 Slug
5 Slugs
Catalog number 035365.QI
also known as 035365-QI
Price (USD)
84.65
Online Exclusive
94.30
Save 9.65 (10%)
Each
Quantity:
1 Slug
Request bulk or custom format
Price (USD)
84.65
Online Exclusive
94.30
Save 9.65 (10%)
Each
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSlug, 6.35mm (0.25in) diameter.
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.99% (metals basis excluding Ta), Ta 300ppm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only