Dionex™ IonPac™ AS7 IC Columns
Actual product may vary
Dionex™ IonPac™ AS7 IC Columns
Dionex™ IonPac™ AS7 IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS7 IC Columns

Green features
Separate of a wide variety of polyvalent anions, including hexavalent chromium by U.S. EPA Method 218.6, using this hydrophobic hydroxide-selective anion-exchange column.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
035393250 mm4 mmAnalytical Column
063097250 mm2 mmAnalytical Column
03539450 mm4 mmGuard Column
06309950 mm2 mmGuard Column
Catalog number 035393
Price (USD)
1,930.00
Each
Length (Metric):
250 mm
Diameter (Metric):
4 mm
Column Type:
Analytical Column
Price (USD)
1,930.00
Each
For separation of a wide variety of polyvalent anions, including polyphosphates, polyphosphonates, and other polyvalent complexing agents, the Thermo Scientific™ Dionex™ IonPac™ AS7 anion-exchange column delivers excellent results. The column has a unique polymer packing that provides superior performance for separating ionic and polar compounds that offers high speed, high efficiency, and high loading capacity at moderate backpressures. However, the newer Dionex IonPac AS16 column is recommended for determination of polyphosphates.
Analysis of Polyvalent Anions and Chelating Agents in Diverse Matrices
  • Analyze hexavalent chromium in environmental matrices in accordance with U.S. EPA Method 218.6
  • For separation of polyvalent anions and chelating agents in complex sample matrices using a post column reagent and UV detection
  • Determines cyanide, sulfide and iodide in high ionic strength matrices using amperometric detection
  • Typically used with post column reagents for photometric detection
  • Contains both anionic and cationic ion-exchange capacity
  • Compatible with a wide range of eluents and detectors for sensitive and selective analysis
Multiple Formats Available for Application Flexibility
  • 4 x 250mm, 2 x 250mm analytical columns
  • 4 x 50mm, 2 x 50mm guard columns
  • Longer 250mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceU.S. EPA Method 218.6
Column Capacity100 μeq
Column ChemistryAcidic
Column ConstructionPEEK
Crosslinking5% DVB
Flow Rate0.5 to 3.0 mL/min.
For Use With (Application)Separation of Polyvalent Anions and Chelating Agents in Complex Sample Matrices
For Use With (Equipment)Eluent and Detector
Green FeaturesFSC
HydrophobicityMedium-High
Ion Exchange GroupAlkyl Quaternary Ammonium
Max. Pressure4000 psi (275 bar)
Particle Size10 μm
QuantityEach 1
Solvent CompatibilitypH 0 to 14
Stationary PhaseIonPac AS7
Column TypeAnalytical Column
Diameter (Metric)4 mm
Length (Metric)250 mm
pH0 to 14
Unit SizeEach