Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL
Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL
Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL
Thermo Scientific Chemicals

Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL

CAS: 7558-80-7 | H2NaO4P | 119.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 035548.AP
also known as 035548-AP
Price (USD)
230.65
Online Exclusive
256.00
Save 25.35 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
230.65
Online Exclusive
256.00
Save 25.35 (10%)
Each
Specifications
CAS7558-80-7
ChEBICHEBI:37585
Chemical Name or MaterialPhosphate
IUPAC Namesodium;dihydrogen phosphate
InChI KeyAJPJDKMHJJGVTQ-UHFFFAOYSA-M
View more
Phosphate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only