Bromide, Ion chromatography standard solution, Specpure™ Br∣- 1000μg/mL
Thermo Scientific Chemicals

Bromide, Ion chromatography standard solution, Specpure™ Br∣- 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 035552.AE
also known as 035552-AE
Price (USD)
128.65
Online Exclusive
143.00
Save 14.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
128.65
Online Exclusive
143.00
Save 14.35 (10%)
Each
Specifications
Chemical Name or MaterialBromide Ion chromatography standard solution
MDL NumberMFCD00134172
Name NoteBr- 1000μg/mL
Physical FormLiquid
Quantity100 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only