Multi Ion anion IC standard solution, Specpure™ (Dionex) in H{2}O
Thermo Scientific Chemicals

Multi Ion anion IC standard solution, Specpure™ (Dionex) in H{2}O

 
Have Questions?
Catalog NumberQuantity
035565.AE
also known as 035565-AE
100 mL
Catalog number 035565.AE
also known as 035565-AE
Price (USD)
254.65
Online Exclusive
283.00
Save 28.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
254.65
Online Exclusive
283.00
Save 28.35 (10%)
Each
Specifications
Chemical Name or MaterialMulti Ion anion IC standard solution in H2O
EINECS Number231-791-2
MDL NumberMFCD00144813
OdorOdorless
Physical FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only