Aluminum chloride, 6.0N Standardized Solution
Aluminum chloride, 6.0N Standardized Solution
Aluminum chloride, 6.0N Standardized Solution
Thermo Scientific Chemicals

Aluminum chloride, 6.0N Standardized Solution

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number 035568.AE
also known as 035568-AE
Price (USD)
72.50
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
72.50
Each
Specifications
CAS7446-70-0
Chemical Name or MaterialAluminum chloride
ColorColorless
DOT InformationDOT Class: 8; Packing Group: III
Density1.3g/cm3 at 20°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only