Ammonium thiocyanate, 0.1N Standardized Solution
Ammonium thiocyanate, 0.1N Standardized Solution
Ammonium thiocyanate, 0.1N Standardized Solution
Thermo Scientific Chemicals

Ammonium thiocyanate, 0.1N Standardized Solution

CAS: 7732-18-5 | NH4SCN
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number 035580.K2
also known as 035580-K2
Price (USD)
61.70
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
61.70
Each
Specifications
CAS7732-18-5
Chemical Name or MaterialAmmonium thiocyanate
ColorColorless
Density1g/cm3 at 20°C
EINECS Number217-175-6
View more
Ammonium thiocyanate, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only