Potassium permanganate, 0.1N Standardized Solution
Potassium permanganate, 0.1N Standardized Solution
Potassium permanganate, 0.1N Standardized Solution
Thermo Scientific Chemicals

Potassium permanganate, 0.1N Standardized Solution

CAS: 7722-64-7 | KMnO4 | 158.032 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035590.K7
also known as 035590-K7
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Chemical Identifiers
CAS7722-64-7
IUPAC Namepotassium manganesoylolate
Molecular FormulaKMnO4
InChI KeyVZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES[K+].[O-][Mn](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration0.1N Potassium permanganate
Potassium permanganate, 0.1N standardized solution is used for volumetric analysis in laboratories. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only