Silver nitrate, 0.1N Standardized Solution
Silver nitrate, 0.1N Standardized Solution
Silver nitrate, 0.1N Standardized Solution
Silver nitrate, 0.1N Standardized Solution
Thermo Scientific Chemicals

Silver nitrate, 0.1N Standardized Solution

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
20 L
Catalog number 035601.K2
also known as 035601-K2
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7761-88-8
IUPAC Namesilver(1+) nitrate
Molecular FormulaAgNO3
InChI KeySQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES[Ag+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
FormLiquid
Concentration0.100N ± 0.14%
Silver nitrate, 1.0N Standardized Solution is used for volumetric analysis in laboratories. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only