Hydrochloric acid, 50% v/v aq. soln.
Hydrochloric acid, 50% v/v aq. soln.
Hydrochloric acid, 50% v/v aq. soln.
Thermo Scientific Chemicals

Hydrochloric acid, 50% v/v aq. soln.

CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035607.K7
also known as 035607-K7
Price (USD)
101.65
Special offer
Online exclusive
Ends: 30-Jun-2026
119.00
Save 17.35 (15%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
101.65
Special offer
Online exclusive
Ends: 30-Jun-2026
119.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
FormLiquid
Assay from Supplier's CofA50% w/v
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only