Lactic acid, 1.0N Standardized Solution
Lactic acid, 1.0N Standardized Solution
Lactic acid, 1.0N Standardized Solution
Thermo Scientific Chemicals

Lactic acid, 1.0N Standardized Solution

CAS: 50-21-5 | C3H6O3 | 90.08 g/mol
Have Questions?
Catalog NumberQuantity
035628.AP
also known as 035628-AP
500 mL
Catalog number 035628.AP
also known as 035628-AP
Price (USD)
104.00
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
104.00
Each
Chemical Identifiers
CAS50-21-5
IUPAC Name2-hydroxypropanoic acid
Molecular FormulaC3H6O3
InChI KeyJVTAAEKCZFNVCJ-UHFFFAOYNA-N
SMILESCC(O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Concentration1.00N ± 0.21%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only