Sodium hydroxide, 2.0N Standardized Solution
Sodium hydroxide, 2.0N Standardized Solution
Sodium hydroxide, 2.0N Standardized Solution
Thermo Scientific Chemicals

Sodium hydroxide, 2.0N Standardized Solution

CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number 035631.K7
also known as 035631-K7
Price (USD)
81.65
Special offer
Online exclusive
Ends: 30-Jun-2026
96.20
Save 14.55 (15%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
81.65
Special offer
Online exclusive
Ends: 30-Jun-2026
96.20
Save 14.55 (15%)
Each
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay from Suppliers CofA2.0N ± 0.24%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only