Iodine, 0.01N Standardized Solution
Iodine, 0.01N Standardized Solution
Iodine, 0.01N Standardized Solution
Thermo Scientific Chemicals

Iodine, 0.01N Standardized Solution

CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035636.K2
also known as 035636-K2
Price (USD)
60.90
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
60.90
Each
Chemical Identifiers
CAS7553-56-2
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Dark brown
Assay from Suppliers CofA0.0100N (±0.29%)
Iodine, 0.01N standardized solution is used for accurate volumetric analysis. It is also used as a disinfectant and antibacterial agent in solutions, tinctures and salves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only