Nickel sulfide, 99.9% (metals basis)
Nickel sulfide, 99.9% (metals basis)
Nickel sulfide, 99.9% (metals basis)
Thermo Scientific Chemicals

Nickel sulfide, 99.9% (metals basis)

CAS: 12035-72-2 | Ni3S2 | 240.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 035661.18
also known as 035661-18
Price (USD)
493.65
Special offer
Online exclusive
Ends: 30-Jun-2026
580.00
Save 86.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
493.65
Special offer
Online exclusive
Ends: 30-Jun-2026
580.00
Save 86.35 (15%)
Each
Chemical Identifiers
CAS12035-72-2
IUPAC Namedisulfanylidenetrinickel
Molecular FormulaNi3S2
InChI KeyYGHCWPXPAHSSNA-UHFFFAOYSA-N
SMILESS=[Ni][Ni][Ni]=S
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only