Praseodymium(III) oxide, 99.9% (metals basis)
Praseodymium(III) oxide, 99.9% (metals basis)
Praseodymium(III) oxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Praseodymium(III) oxide, 99.9% (metals basis)

CAS: 12036-32-7 | O3Pr2 | 329.812 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 035663.09
also known as 035663-09
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
134.00
Save 20.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
134.00
Save 20.35 (15%)
Each
Chemical Identifiers
CAS12036-32-7
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Praseodymium(III) oxide is used as a pigment in glass, optic and ceramic industries. Praseodymium-doped glass called didymium glass is used in welding goggles, since it blocks infra-red radiation. Further, it is used as a dielectric in combination with silicon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only