Calcium tungsten oxide, 99.78% (metals basis)
Calcium tungsten oxide, 99.78% (metals basis)
Calcium tungsten oxide, 99.78% (metals basis)
Thermo Scientific Chemicals

Calcium tungsten oxide, 99.78% (metals basis)

CAS: 7790-75-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 035669.18
also known as 035669-18
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
106.00
Save 15.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
106.00
Save 15.35
Each
Chemical Identifiers
CAS7790-75-2
Molecular FormulaCaO4W
InChI KeyFDMFQOCGNBYKPY-UHFFFAOYSA-N
SMILES[Ca++].[O-][W]([O-])(=O)=O
Molecular Weight (g/mol)287.91
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.22% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only