Copper(II) chloride, ultra dry, 99.995% (metals basis)
Copper(II) chloride, ultra dry, 99.995% (metals basis)
Copper(II) chloride, ultra dry, 99.995% (metals basis)
Thermo Scientific Chemicals

Copper(II) chloride, ultra dry, 99.995% (metals basis)

Air Sensitive and Hygroscopic. >300°C partial decomposition to CuCl and Cl2 | CAS: 7447-39-4 | CuCl2
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 035673.06
also known as 035673-06
Price (USD)
71.65
Online Exclusive
79.50
Save 7.85 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
71.65
Online Exclusive
79.50
Save 7.85 (10%)
Each
Chemical Identifiers
CAS7447-39-4
IUPAC Namecopper(2+) dichloride
Molecular FormulaCl2Cu
InChI KeyORTQZVOHEJQUHG-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Cu++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
FormPowder
Residual water< 100 ppm
Total Metal Impurities0.005% max.
Copper(II) chloride is used as a co-catalyst with palladium(II) chloride used in the Wacker process to prepare ethanol from ethane. It acts as an effective catalyst for the tetrahydropyranylation of alcohols. It is involved in the production of vinyl chloride and dichloroethane as well as in the chlorination of aromatic hydrocarbons. It acts as a deodorizing and desulfurizing agent in petroleum industry, mordant in textiles industry and in electrotyping baths for plating copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only